accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH2C(R)O(S)(R)CH3(S)HO(R)(S)OH3CCH3(S)(R)CH2ClHNHO(S)CH3(R)O(S)(S)HO

CAS RN 12627-35-9

IUPAC Name: (1S,2R,5S,6R,8S,9S,10R,12S,15R,16S,25R,27S)-21-chloro-15,16,33,33-tetramethyl-24-methylidene-10-(prop-1-en-2-yl)-7,11,32-trioxa-18-azadecacyclo[25.4.2.0²,¹⁶.0⁵,¹⁵.0⁶,⁸.0⁶,¹².0¹⁷,³¹.0¹⁹,³⁰.0²²,²⁹.0²⁵,²⁸]tritriaconta-17(31),19,21,29-tetraene-5,9,28-triol
Synonyms: penitrem A
Molecular Weight (g/mol): 634.21
Molecular Formula: C37H44ClNO6
InChi Key: JDUWHZOLEDOQSR-JHMXYHNCSA-N
SMILES: CC(=C)[C@H]1O[C@H]2CC[C@]3(C)[C@]4(C)[C@@H](CC[C@@]3(O)[C@@]22O[C@H]2[C@H]1O)[C@@H]1OC(C)(C)[C@H]2C[C@@H]3C(=C)CC5=C(Cl)C=C6NC4=C1C6=C5C23O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results