CAS RN 126403-67-6
IUPAC Name:
2-methylpropan-1-ide; bromozincylium
Synonyms:
2-methylpropan-1-ide; bromozincylium
Molecular Weight (g/mol):
202.4
Molecular Formula:
C4H9BrZn
InChi Key:
DSDCDMKASWVZHI-UHFFFAOYSA-M
SMILES:
[Zn+]Br.CC(C)[CH2-]
1
–
1
of
1
results
1
Sigma Aldrich 3-Cyanopropylzinc Bromide Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.