accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OBOHHO

CAS RN 126617-98-9

IUPAC Name: [2-(methoxymethyl)phenyl]boronic acid
Synonyms: 2-(methoxymethyl)phenylboronic acid
Molecular Weight (g/mol): 165.98
Molecular Formula: C8H11BO3
InChi Key: JHPPAFXQJZJGEP-UHFFFAOYSA-N
SMILES: COCC1=CC=CC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results