accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOClClONa

CAS RN 1266615-56-8

IUPAC Name: sodium 4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
Synonyms: 2,6-dichloroindophenol sodium
Molecular Weight (g/mol): 290.07
Molecular Formula: C12H6Cl2NNaO2
InChi Key: CVSUAFOWIXUYQA-UHFFFAOYSA-M
SMILES: [Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1
Molecular Weight (g/mol) 
  • (10)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (3)
  • (4)
Percent Purity 
  • (4)
  • (3)
Color 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (1)
  • (2)
  • (10)
  • (10)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (4)
  • (4)
  • (3)
  • (1)
  • (4)