Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloroindophenol Sodium Salt Hydrate, Sigma-Aldrich

Catalog No. 01258810 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-810 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-810 Supplier MilliporeSigma Supelco Supplier No. D18785G
Add to Cart
Add to Cart
  • 2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. 2,6-Dichloroindophenol (DCIP) may be used in the following studies:
  • As redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for enzymatic activity of glucose oxidase (GOx).
  • To compose UVB specific dosimeter.
  • Quantification of vitamin C (L-ascorbic acid) in fruit juices.
  • As substrate in NQO1 assay to investigate the liver cytosolic NQO1 activity spectrophotometrically.
  • Mitochondrial succinate CoQ oxidoreductase (complex II) activity assay.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1266615-56-8
Molecular Weight (g/mol) 290.08 g/mol

Specifications

Color Dark Blue
Quantity 5 g
Physical Form Powder
Chemical Name or Material 2,6-Dichloroindophenol sodium salt hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.