accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOHO

CAS RN 1266615-78-4

IUPAC Name: 3-formyl-2-hydroxybenzoic acid
Synonyms: 3-formylsalicylic acid
Molecular Weight (g/mol): 166.13
Molecular Formula: C8H6O4
InChi Key: YOEUNKPREOJHBW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(C=O)=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)