accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1266615-79-5

ONBOHOHH3C

CAS RN 1266615-79-5

IUPAC Name: (4-methoxypyridin-3-yl)boronic acid
Synonyms: 4-methoxypyridin-3-ylboronic acid
Molecular Weight (g/mol): 152.94
Molecular Formula: C6H8BNO3
InChi Key: YUTPAZKVEOJQCY-UHFFFAOYSA-N
SMILES: COC1=CC=NC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results