CAS RN 126705-22-4
CAS RN 126705-22-4
IUPAC Name:
1-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)cyclopropane-1-carboxylic acid
Synonyms:
1-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}cyclopropane-1-carboxylic acid
Molecular Weight (g/mol):
323.35
Molecular Formula:
C19H17NO4
InChi Key:
OPPOISJKHBLNPD-UHFFFAOYSA-N
SMILES:
OC(=O)C1(CC1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-ACPC-OH, Advanced ChemTech
CAS: 126705-22-4 Molecular Formula: C19H17NO4 Molecular Weight (g/mol): 323.35 MDL Number: MFCD00190873 InChI Key: OPPOISJKHBLNPD-UHFFFAOYSA-N IUPAC Name: 1-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)cyclopropane-1-carboxylic acid SMILES: OC(=O)C1(CC1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2