accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHOOOOOOOOHOSOOOOOOOOOOHOSOOOOOOOHOOHOHHNOCH3OHOHHNOCH3OHOHOSOOOOHOH

CAS RN 12678-07-8

IUPAC Name: 6-[(2-{[2-carboxylato-6-({2-[(2-carboxylato-4,5,6-trihydroxyoxan-3-yl)oxy]-3-acetamido-5-hydroxy-6-[(sulfonatooxy)methyl]oxan-4-yl}oxy)-4,5-dihydroxyoxan-3-yl]oxy}-3-acetamido-5-hydroxy-6-[(sulfonatooxy)methyl]oxan-4-yl)oxy]-3-({3-acetamido-4,5-dihydroxy-6-[(sulfonatooxy)methyl]oxan-2-yl}oxy)-4,5-dihydroxyoxane-2-carboxylate
Synonyms: 6-[(2-{[2-carboxylato-6-({2-[(2-carboxylato-4,5,6-trihydroxyoxan-3-yl)oxy]-3-acetamido-5-hydroxy-6-[(sulfonatooxy)methyl]oxan-4-yl}oxy)-4,5-dihydroxyoxan-3-yl]oxy}-3-acetamido-5-hydroxy-6-[(sulfonatooxy)methyl]oxan-4-yl)oxy]-3-({3-acetamido-4,5-dihydroxy-6-[(sulfonatooxy)methyl]oxan-2-yl}oxy)-4,5-dihydroxyoxane-2-carboxylate
Molecular Weight (g/mol): 1390.1
Molecular Formula: C42H59N3O43S3
InChi Key: ABHQVWCLVQDEKE-UHFFFAOYNA-H
SMILES: CC(=O)NC1C(O)C(O)C(COS([O-])(=O)=O)OC1OC1C(O)C(O)C(OC2C(O)C(COS([O-])(=O)=O)OC(OC3C(O)C(O)C(OC4C(O)C(COS([O-])(=O)=O)OC(OC5C(O)C(O)C(O)OC5C([O-])=O)C4NC(C)=O)OC3C([O-])=O)C2NC(C)=O)OC1C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results