Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chondroitin Sulfate C, MP Biomedicals

Catalog No. ICN970551 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN970551 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN970551 Supplier MP Biomedicals, Inc Supplier No. 08970551

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

From Shark Cartilage Sodium Salt Inorganic sulfur and chloride are not detected. The Mrin 4-8 x 104and the 4 to 6 sulfate ratio is 10:90

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12678-07-8
Molecular Formula C42H59N3O43S3-6
Molecular Weight (g/mol) 1390.092
InChI Key ABHQVWCLVQDEKE-UHFFFAOYSA-H
Synonym yl-1->4 hexopyranuronate, chondroitin 6-sulfate sodium salt?, 2-acetamido-2-deoxy-6-o-sulfonatohexopyranosyl-1->4 hexopyranuronosyl-1->3-2-acetamido-2-deoxy-6-o-sulfonatohexopyranosyl-1->4 hexopyranuronosyl-1->3-2-acetamido-2-deoxy-6-o-sulfonatohexopyranos, 3-3-acetamido-4-5-3-acetamido-4-5-3-acetamido-4,5-dihydroxy-6-sulfonatooxymethyl-2-oxanyl oxy-6-carboxylato-3,4-dihydroxy-2-oxanyl oxy-5-hydroxy-6-sulfonatooxymethyl-2-oxanyl oxy-6-carboxylato-3,4-dihydroxy-2-oxanyl oxy-5-hydroxy-6-sulf, 3-3-acetamido-4-5-3-acetamido-4-5-3-acetamido-4,5-bis oxidanyl-6-sulfonatooxymethyl oxan-2-yl oxy-6-carboxylato-3,4-bis oxidanyl oxan-2-yl oxy-5-oxidanyl-6-sulfonatooxymethyl oxan-2-yl oxy-6-carboxylato-3,4-bis oxidanyl oxan-2-yl oxy-5-oxidanyl-6, 6-2-2-carboxylato-6-2-2-carboxylato-4,5,6-trihydroxyoxan-3-yl oxy-3-acetamido-5-hydroxy-6-sulfonatooxy methyl oxan-4-yl oxy-4,5-dihydroxyoxan-3-yl oxy-3-acetamido-5-hydroxy-6-sulfonatooxy methyl oxan-4-yl oxy-3-3-acetamido-4,5-dihydroxy-6-sulfonatooxy met
PubChem CID 42609007
IUPAC Name 3-[3-acetamido-4-[5-[3-acetamido-4-[5-[3-acetamido-4,5-dihydroxy-6-(sulfonatooxymethyl)oxan-2-yl]oxy-6-carboxylato-3,4-dihydroxyoxan-2-yl]oxy-5-hydroxy-6-(sulfonatooxymethyl)oxan-2-yl]oxy-6-carboxylato-3,4-dihydroxyoxan-2-yl]oxy-5-hydroxy-6-(sulfonatooxym
SMILES CC(=O)NC1C(C(C(OC1OC2C(C(C(OC2C(=O)[O-])OC3C(C(OC(C3O)COS(=O)(=O)[O-])OC4C(C(C(OC4C(=O)[O-])OC5C(C(OC(C5O)COS(=O)(=O)[O-])OC6C(C(C(OC6C(=O)[O-])O)O)O)NC(=O)C)O)O)NC(=O)C)O)O)COS(=O)(=O)[O-])O)O

Specifications

Quantity 10 mg
Chemical Name or Material Chondroitin Sulfate C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.