CAS RN 126850-04-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Supelco™ Diol-Functionalized Silica Gel, Spherical, 40-70 μm
Diol-functionalized silica gel may be used as stationary phase for liquid chromatographic analysis.
2
MilliporeSigma™ Supelco™ Diol-Functionalized Silica Gel, Spherical, 40-70 μm
Diol-functionalized silica gel may be used as stationary phase for liquid chromatographic analysis.