Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Diol-Functionalized Silica Gel, Spherical, 40-70 μm

Catalog No. 111012881 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2881 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2881 Supplier MilliporeSigma™ Supelco™ Supplier No. 939811KG
Add to Cart
Add to Cart

Diol-functionalized silica gel may be used as stationary phase for liquid chromatographic analysis.

Diol-functionalized silica gel is less polar and has lower retention time when compared to normal phase silica gel.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 40 to 70 μm
Pore Size 110 Å
Grade Standard
Quantity 1 kg
Surface Area (Metric) ˜300 m2/g
Description Carbon content, 4.0 to 7.0%
Physical Form Powder
CAS 126850-04-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.