accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiH3CH3CCH3OSOOFFF

CAS RN 126889-55-2

IUPAC Name: diethyl(propan-2-yl)silyl trifluoromethanesulfonate
Synonyms: diethyl(isopropyl)silyl trifluoromethanesulfonate
Molecular Weight (g/mol): 278.36
Molecular Formula: C8H17F3O3SSi
InChi Key: RIXRWPWLOMOUPT-UHFFFAOYSA-N
SMILES: CC[Si](CC)(OS(=O)(=O)C(F)(F)F)C(C)C
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results