CAS RN 127-09-3
Sodium acetate, anhydrous, 99%
CAS: 127-09-3 Molecular Formula: C2H3NaO2 Molecular Weight (g/mol): 82.03 MDL Number: MFCD00012459 InChI Key: VMHLLURERBWHNL-UHFFFAOYSA-M Synonym: sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech PubChem CID: 517045 ChEBI: CHEBI:32954 SMILES: [Na+].CC([O-])=O
Sodium acetate, anhydrous, ACS, 99.0% min
CAS: 127-09-3 Molecular Formula: C2H3NaO2 Molecular Weight (g/mol): 82.03 MDL Number: MFCD00012459 InChI Key: VMHLLURERBWHNL-UHFFFAOYSA-M Synonym: sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech PubChem CID: 517045 ChEBI: CHEBI:32954 IUPAC Name: sodium acetate SMILES: [Na+].CC([O-])=O
Sodium acetate, 3M aq. soln., pH 5.2, RNAse free
CAS: 127-09-3 Molecular Formula: C2H3NaO2 Molecular Weight (g/mol): 82.03 MDL Number: MFCD00012459 InChI Key: VMHLLURERBWHNL-UHFFFAOYSA-M Synonym: sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech PubChem CID: 517045 ChEBI: CHEBI:32954 IUPAC Name: sodium acetate SMILES: [Na+].CC([O-])=O
Sodium acetate, anhydrous, 99.997% (metals basis)
CAS: 127-09-3 Molecular Formula: C2H3NaO2 Molecular Weight (g/mol): 82.03 MDL Number: MFCD00012459 InChI Key: VMHLLURERBWHNL-UHFFFAOYSA-M Synonym: sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech PubChem CID: 517045 ChEBI: CHEBI:32954 IUPAC Name: sodium acetate SMILES: [Na+].CC([O-])=O
Sodium Acetate Anhydrous (Fused Crystals/Certified ACS), Fisher Chemical™
CAS: 127-09-3 Molecular Formula: C2H3NaO2 Molecular Weight (g/mol): 82.03 MDL Number: MFCD00012459 InChI Key: VMHLLURERBWHNL-UHFFFAOYSA-M Synonym: sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech PubChem CID: 517045 ChEBI: CHEBI:32954 IUPAC Name: sodium acetate SMILES: [Na+].CC([O-])=O
Sodium Acetate Anhydrous (White Crystals or Granular Powder), Fisher BioReagents
CAS: 127-09-3 Molecular Formula: C2H3NaO2 Molecular Weight (g/mol): 82.03 MDL Number: MFCD00012459 InChI Key: VMHLLURERBWHNL-UHFFFAOYSA-M Synonym: sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech PubChem CID: 517045 ChEBI: CHEBI:32954 SMILES: [Na+].CC([O-])=O
Thermo Scientific Chemicals Acetate Buffer (1 M, pH 4.0)
CAS: 127-09-3
Sodium acetate, 3M aq. soln., pH 4.5, autoclaved
CAS: 127-09-3 Molecular Formula: C2H3NaO2 Molecular Weight (g/mol): 82.03 MDL Number: MFCD00012459 InChI Key: VMHLLURERBWHNL-UHFFFAOYSA-M Synonym: sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech PubChem CID: 517045 ChEBI: CHEBI:32954 SMILES: [Na+].CC([O-])=O
Sodium acetate, 1M aq. soln., pH 4.5, RNAse free
CAS: 127-09-3 Molecular Formula: C2H3NaO2 Molecular Weight (g/mol): 82.03 MDL Number: MFCD00012459 InChI Key: VMHLLURERBWHNL-UHFFFAOYSA-M Synonym: sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech PubChem CID: 517045 ChEBI: CHEBI:32954 SMILES: [Na+].CC([O-])=O
Acetic acid, sodium salt, 99%, for HPLC, anhydrous
CAS: 127-09-3 Molecular Formula: C2H3NaO2 Molecular Weight (g/mol): 82.03 MDL Number: MFCD00012459 InChI Key: VMHLLURERBWHNL-UHFFFAOYSA-M Synonym: sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech PubChem CID: 517045 ChEBI: CHEBI:32954 SMILES: [Na+].CC([O-])=O
Thermo Scientific Chemicals Acetate Buffer (1 M, pH 3.5)
CAS: 127-09-3
Sodium acetate, 99+%, anhydrous, Molecular Biology Reagent, Thermo Scientific Chemicals
CAS: 127-09-3 Molecular Formula: C2H3NaO2 Molecular Weight (g/mol): 82.03 MDL Number: MFCD00012459 InChI Key: VMHLLURERBWHNL-UHFFFAOYSA-M Synonym: sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech PubChem CID: 517045 ChEBI: CHEBI:32954 SMILES: [Na+].CC([O-])=O
Acetic acid, sodium salt trihydrate, 99.5%, for analysis
CAS: 127-09-3 Molecular Formula: C2H3NaO2 Molecular Weight (g/mol): 82.03 MDL Number: MFCD00012459 InChI Key: VMHLLURERBWHNL-UHFFFAOYSA-M Synonym: sodium acetate,acetic acid, sodium salt,sodium acetate anhydrous,sodium acetate, anhydrous,acetic acid sodium salt,anhydrous sodium acetate,sodii acetas,sodium ethanoate,natrium aceticum,octan sodny czech PubChem CID: 517045 ChEBI: CHEBI:32954 IUPAC Name: sodium acetate SMILES: [Na+].CC([O-])=O
Thermo Scientific Chemicals Acetate Buffer (1 M, pH 3.0)
CAS: 127-09-3
Thermo Scientific Chemicals Acetate Buffer (1 M, pH 5.5)
CAS: 127-09-3