accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 127-58-2

IUPAC Name: sodium 4-{[(4-methylpyrimidin-2-yl)azanidyl]sulfonyl}aniline
Synonyms: sodium 4-[(4-methylpyrimidin-2-yl)azanidylsulfonyl]aniline
Molecular Weight (g/mol): 286.28
Molecular Formula: C11H11N4NaO2S
InChi Key: BSFJGCCAXDCMOX-UHFFFAOYSA-N
SMILES: [Na+].CC1=CC=NC([N-]S(=O)(=O)C2=CC=C(N)C=C2)=N1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results