Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfamerazine sodium salt

Catalog No. AAJ6671730
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ6671730 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6671730 Supplier Thermo Scientific Chemicals Supplier No. J6671730
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 127-58-2 | C11H11N4NaO2S | 286.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-58-2
Molecular Formula C11H11N4NaO2S
Molecular Weight (g/mol) 286.285
MDL Number MFCD00068332
InChI Key BSFJGCCAXDCMOX-UHFFFAOYSA-N
Synonym sulfamerazine sodium, solumedine, sulfamerazine sodium salt, sodium sulfamerazine, sulfamerazin natrium, sodium sulphamerazine, soluble sulfamerazine, unii-jov4ujy07o, solfamerazina sodica dcit
PubChem CID 15899899
IUPAC Name sodium;(4-aminophenyl)sulfonyl-(4-methylpyrimidin-2-yl)azanide
SMILES CC1=NC(=NC=C1)[N-]S(=O)(=O)C2=CC=C(C=C2)N.[Na+]

Specifications

Packaging Plastic bottle
Quantity 250 g
Solubility Information Soluble in DMSO or water
Formula Weight 286.26
Physical Form Powder
Chemical Name or Material Sulfamerazine sodium salt
Hazard Category H302+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 204-851-0
RTECSNumber WP0760000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.