accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOON

CAS RN 1270976-78-7

IUPAC Name: 3-[(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl]benzonitrile
Synonyms: 3-[(2,5-dioxopyrrol-1-yl)methyl]benzonitrile
Molecular Weight (g/mol): 212.21
Molecular Formula: C12H8N2O2
InChi Key: YQNWHSSASIYIKH-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1CC1=CC=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results