CAS RN 1271-48-3
IUPAC Name:
1,1'-Ferrocenedicarboxaldehyde
Synonyms:
—
Molecular Weight (g/mol):
242.06
Molecular Formula:
C12H10FeO2
InChi Key:
—
SMILES:
[Fe].O=Cc1cccc1.O=Cc1cccc1
1
–
1
of
1
results
1
Sigma Aldrich Gly-His Hydrochloride Hydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.