accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1271-54-1

IUPAC Name: bis(benzene)chromium
Synonyms: bis(benzene)chromium
Molecular Weight (g/mol): 202.18
Molecular Formula: C12H6Cr
InChi Key: WELDVOWFBCCDNE-UHFFFAOYSA-N
SMILES: C1=CC=CC=C1.[C-]12[C-]3[C-]4[C-]5[C-]6[C-]1[Cr]23456

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results