accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeNH3CCH3

CAS RN 1271-86-9

IUPAC Name: (Dimethylaminomethyl)ferrocene
Synonyms:
Molecular Weight (g/mol): 243.13
Molecular Formula: C13H17FeN
InChi Key:
SMILES: [Fe].c1cccc1.CN(C)Cc1cccc1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)