Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Dimethylaminomethyl)ferrocene, 98+%, Thermo Scientific™

Catalog No. AAL0791514
Change view
Click to view available options
Quantity:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0791514 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0791514 Supplier Thermo Scientific Chemicals Supplier No. L0791514

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(Dimethylaminomethyl)ferrocene is used as an active pharmaceutical ingredient.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1271-86-9
Density 1.228
Boiling Point 122°C to 124°C (2mmHg)
Flash Point >110°C (230°F)
Molecular Formula C13H17FeN
Refractive Index 1.591
Quantity 25g
MDL Number MFCD00001433
Solubility Information Difficult to mix.
Formula Weight 243.13
Percent Purity ≥98%
Chemical Name or Material (Dimethylaminomethyl)ferrocene
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 215-044-8
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.