accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 127103-11-1

IUPAC Name: 1-{6-amino-2-[3-carboxy-2-(2-acetamido-3-hydroxypropanamido)propanamido]hexanoyl}pyrrolidine-2-carboxylic acid
Synonyms: 1-{6-amino-2-[3-carboxy-2-(2-acetamido-3-hydroxypropanamido)propanamido]hexanoyl}pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 487.51
Molecular Formula: C20H33N5O9
InChi Key: HJDRXEQUFWLOGJ-UHFFFAOYNA-N
SMILES: CC(=O)NC(CO)C(=O)NC(CC(O)=O)C(=O)NC(CCCCN)C(=O)N1CCCC1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)