Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals N-Acetyl-Ser-Asp-Lys-Pro

Catalog No. AAJ66430LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66430LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66430LB0 Supplier Thermo Scientific Chemicals Supplier No. J66430LB0
Add to Cart

missing translation for 'validMainframeMsg'

 

N-Acetyl-Ser-Asp-Lys-Pro inhibits the entry of pluripotent hemopoietic stem cells into S-phase of the cell cycle and protects against Ara-C lethality in mice. It is a specific substrate for the N-terminal active site of angiotensin converting enzyme, which is responsible for its degradation in vivo.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-Ser-Asp-Lys-Pro inhibits the entry of pluripotent hemopoietic stem cells into S-phase of the cell cycle and protects against Ara-C lethality in mice. It is a specific substrate for the N-terminal active site of angiotensin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127103-11-1

Specifications

Color White
Content And Storage Store at -20°C
Format Solid
Molecular Weight (g/mol) 487.51
Product Type N-Acetyl-Ser-Asp-Lys-Pro
Quantity 5 mg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.