accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeOH

CAS RN 1273-86-5

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 216.06
Molecular Formula: C11H12FeO
InChi Key:
SMILES: [Fe].c1cccc1.OCc1cccc1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)