Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxymethylferrocene 95.0+%, TCI America™
SDP

Catalog No. H04645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04645G 5 g
H04641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04645G Supplier TCI America Supplier No. H04645G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1273-86-5
Molecular Formula C11H12FeO
Molecular Weight (g/mol) 216.06
MDL Number MFCD00040878
InChI Key KLMBXXGIFRROFL-UHFFFAOYSA-N
Synonym Ferrocenylmethanol
SMILES [Fe].c1cccc1.OCc1cccc1

Specifications

Melting Point 78°C
Quantity 5 g
Formula Weight 216.06
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hydroxymethylferrocene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.