accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 127346-48-9

IUPAC Name: tert-butyl N-(3-aminopropyl)carbamate hydrochloride
Synonyms: tert-butyl N-(3-aminopropyl)carbamate hydrochloride
Molecular Weight (g/mol): 210.7
Molecular Formula: C8H19ClN2O2
InChi Key: WUXOJNUZYOFBMI-UHFFFAOYSA-N
SMILES: Cl.CC(C)(C)OC(=O)NCCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)