Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-1,3-diaminopropane hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. AAL1948603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1948603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1948603 Supplier Thermo Scientific Chemicals Supplier No. L1948603
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 127346-48-9 | Molecular Formula: C8H19ClN2O2 | Molecular Weight: 210.702 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127346-48-9
Molecular Formula C8H19ClN2O2
Molecular Weight (g/mol) 210.702
MDL Number MFCD01076598
InChI Key WUXOJNUZYOFBMI-UHFFFAOYSA-N
Synonym tert-butyl 3-aminopropyl carbamate hydrochloride, n-boc-1,3-diaminopropane hydrochloride, n-3-aminopropyl carbamic acid tert-butyl ester hydrochloride, n-boc-1,3-propanediamine hydrochloride, tert-butyl n-3-aminopropyl carbamate hydrochloride, acmc-20ak6b, n-boc-1,3-diaminopropane hcl, n-3-aminopropyl carbamic acid tert-butyl ester-hcl, tert-butyl 3-aminopropyl carbamatehydrochloride, n-3-aminopropyl carbamic acid tert-butyl ester hcl
PubChem CID 21982912
IUPAC Name tert-butyl N-(3-aminopropyl)carbamate;hydrochloride
SMILES CC(C)(C)OC(=O)NCCCN.Cl

Specifications

Melting Point 159°C to 161°C
Beilstein 3627894
Quantity 1 g
Formula Weight 210.71
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material N-Boc-1,3-diaminopropane hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.