accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3HOHOOOHO

CAS RN 127464-43-1

IUPAC Name: 2-ethyl-6-methylpyridin-3-ol; butanedioic acid
Synonyms: emoxypine; succinic acid
Molecular Weight (g/mol): 255.27
Molecular Formula: C12H17NO5
InChi Key: IKMNOGHPKNFPTK-UHFFFAOYSA-N
SMILES: OC(=O)CCC(O)=O.CCC1=NC(C)=CC=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results