Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-3-hydroxy-6-methylpyridine Succinate 98.0+%, TCI America™
SDP

Catalog No. E10691G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E10691G Supplier TCI America Supplier No. E10691G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antioxidant and membrane-protective agent for research use.

Chemical Identifiers

CAS 127464-43-1
Molecular Formula C12H17NO5
Molecular Weight (g/mol) 255.27
MDL Number MFCD00445612
InChI Key IKMNOGHPKNFPTK-UHFFFAOYSA-N
Synonym 2-Ethyl-6-methyl-3-pyridinol Succinate, Emoxypine Succinate
PubChem CID 122298
IUPAC Name butanedioic acid;2-ethyl-6-methylpyridin-3-ol
SMILES CCC1=C(C=CC(=N1)C)O.C(CC(=O)O)C(=O)O

Specifications

Melting Point 113°C
Color White
Quantity 1 g
Formula Weight 255.27
Percent Purity ≥98.0% (T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethyl-3-hydroxy-6-methylpyridine Succinate
RTECSNumber EJ9967500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.