accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1277-49-2

IUPAC Name: (±)-1-Ferrocenylethanol
Synonyms:
Molecular Weight (g/mol): 230.09
Molecular Formula: C12H14FeO
InChi Key:
SMILES: [Fe].c1cccc1.CC(O)c1cccc1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results