Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hydroxyethylferrocene 95.0+%, TCI America™
SDP

Catalog No. H04635G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04635G 5 g
H04631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04635G Supplier TCI America Supplier No. H04635G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1277-49-2
Molecular Formula C12H14FeO
Molecular Weight (g/mol) 230.09
MDL Number MFCD00013752
InChI Key KGIBJAFJTDOHBV-LURJTMIESA-N
Synonym 1-Ferrocenylethanol
IUPAC Name (±)-1-Ferrocenylethanol
SMILES [Fe].c1cccc1.CC(O)c1cccc1

Specifications

Melting Point 78°C
Quantity 5 g
Formula Weight 230.09
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Hydroxyethylferrocene
RTECSNumber LK0730000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.