CAS RN 127733-98-6
IUPAC Name:
platinum(2+) tetraamine hydrogen phosphate
Synonyms:
platinum(2+) tetraamine hydrogen phosphate
Molecular Weight (g/mol):
359.19
Molecular Formula:
H13N4O4PPt
InChi Key:
LSDWMQXWIQMHNQ-UHFFFAOYSA-L
SMILES:
N.N.N.N.[Pt++].OP([O-])([O-])=O
1
–
1
of
1
results
1
Tetraammineplatinum(II) hydrogen phosphate solution, Pt 0.5% w/w (cont. Pt), Thermo Scientific Chemicals
CAS: 127733-98-6 Molecular Formula: H13N4O4PPt Molecular Weight (g/mol): 359.19 MDL Number: MFCD03788258 InChI Key: LSDWMQXWIQMHNQ-UHFFFAOYSA-L Synonym: azane;hydrogen phosphate;platinum 2+,platinum 2+ tetraamine hydrogen phosphate,platinum 2+ hydrogen phosphate-ammonia 1/1/4,platinum 2+ ion tetraamine hydrogen phosphate IUPAC Name: platinum(2+) tetraamine hydrogen phosphate SMILES: N.N.N.N.[Pt++].OP([O-])([O-])=O