CAS RN 127783-36-2
IUPAC Name:
phenyl[2-(trimethylsilyl)ethynyl]iodanium; tetrafluoroboranuide
Synonyms:
phenyl[2-(trimethylsilyl)ethynyl]iodanium tetrafluoroborate
Molecular Weight (g/mol):
388.03
Molecular Formula:
C11H14BF4ISi
InChi Key:
QHLUVBBDXOGLSV-UHFFFAOYSA-N
SMILES:
F[B-](F)(F)F.C[Si](C)(C)C#C[I+]C1=CC=CC=C1
1
–
1
of
1
results
1
Trimethylsilylethynyl(phenyl)iodonium Tetrafluoroborate 98.0+%, TCI America™
CAS: 127783-36-2 Molecular Formula: C11H14BF4ISi Molecular Weight (g/mol): 388.026 MDL Number: MFCD00191578 InChI Key: QHLUVBBDXOGLSV-UHFFFAOYSA-N Synonym: Phenyl(trimethylsilylethynyl)iodonium Tetrafluoroborate PubChem CID: 10949095 IUPAC Name: phenyl(2-trimethylsilylethynyl)iodanium;tetrafluoroborate SMILES: [B-](F)(F)(F)F.C[Si](C)(C)C#C[I+]C1=CC=CC=C1