Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsilylethynyl(phenyl)iodonium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. P12391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P12391G Supplier TCI America Supplier No. P12391G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 127783-36-2
Molecular Formula C11H14BF4ISi
Molecular Weight (g/mol) 388.026
MDL Number MFCD00191578
InChI Key QHLUVBBDXOGLSV-UHFFFAOYSA-N
Synonym Phenyl(trimethylsilylethynyl)iodonium Tetrafluoroborate
PubChem CID 10949095
IUPAC Name phenyl(2-trimethylsilylethynyl)iodanium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C[Si](C)(C)C#C[I+]C1=CC=CC=C1

Specifications

Quantity 1 g
UN Number 2925
Formula Weight 388.03
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylsilylethynyl(phenyl)iodonium Tetrafluoroborate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.