accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SH3CCH3FFFSOOO

CAS RN 127820-38-6

IUPAC Name: tris(4-methylphenyl)sulfanium trifluoromethanesulfonate
Synonyms: tris(4-methylphenyl)sulfanium triflate
Molecular Weight (g/mol): 454.52
Molecular Formula: C22H21F3O3S2
InChi Key: ZZJNLOGMYQURDL-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.CC1=CC=C(C=C1)[S+](C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results