accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHH3CCH3H3CCH3H3CCH3

CAS RN 128-37-0

IUPAC Name: 2,6-di-tert-butyl-4-methylphenol
Synonyms: ional ionole sustane bht tonarol alkofen BP parabar 441 antioxidant 264 antox QT butylhydroxytoluene hydroxytoluene, butylated
Molecular Weight (g/mol): 220.36
Molecular Formula: C15H24O
InChi Key: NLZUEZXRPGMBCV-UHFFFAOYSA-N
SMILES: CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (12)
  • (11)
  • (9)
Quantity 
  • (5)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (6)
Percent Purity 
  • (7)
  • (3)
  • (10)
  • (1)
  • (4)
Physical Form 
  • (3)
  • (2)
  • (9)
  • (7)
Boiling Point 
  • (2)
  • (2)
  • (5)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (18)
  • (9)
  • (2)
  • (1)
  • (4)
  • (5)
  • (9)
  • (1)
  • (2)
  • (9)
  • (1)
  • (9)
  • (2)
  • (2)
  • (12)
  • (11)
  • (9)
  • (5)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (6)
  • (7)
  • (3)
  • (10)
  • (1)
  • (4)
  • (3)
  • (2)
  • (9)
  • (7)
  • (2)
  • (2)
  • (5)
  • (7)
  • (1)
  • (5)
  • (4)
  • (1)