Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Di-tert-4-butylhydroxytoluene (BHT), MilliporeSigma™ Supelco™

Catalog No. 111020646 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0646 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0646 Supplier MilliporeSigma Supelco Supplier No. 47168
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3,5-Di-tert-4-butylhydroxytoluene belongs to the class of phenolic antioxidants, added to food and other products such as oils, fats and butter oil to prevent rancidity caused by the oxidation of unsaturated fats.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 128-37-0
Flash Point 127°C
Physical Form Neat
MDL Number MFCD00011644
Synonym 2,6-Di-tert-butyl-4-methylphenol (BHT); E321
Type Food Additive Standard
Quantity 500 mg
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.