accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 128-44-9

IUPAC Name: sodium 1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate
Synonyms: sodium 1,3-dioxo-1λ⁶,2-benzothiazol-1-olate
Molecular Weight (g/mol): 205.16
Molecular Formula: C7H4NNaO3S
InChi Key: OAZGZMKGRPRHJX-UHFFFAOYSA-M
SMILES: [Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12
Grade 
  • (1)
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)