Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saccharin Sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012910 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2910 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2910 Supplier MilliporeSigma Supelco Supplier No. PHR1348500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Saccharin Sodium is used as a sweetening and flavoring agent in food products and beverages. It is also used for masking unpleasant taste in pharmaceutical formulations such as tablets and powders. Its sweetening power is approximately 300 times more than sucrose.Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards.
TRUSTED_SUSTAINABILITY

Specifications

CAS 128-44-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C7H4 NNaO3 S
Synonym Saccharin sodium; Saccharin sodium salt
Type Certified Reference Material
Quantity 500 mg
Formula Weight 205.17
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.