accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOO

CAS RN 128-53-0

IUPAC Name: 1-ethyl-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: N-ethylmaleimide NEM maleic acid N-ethylimide nem (n ethylmaleimide)
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: HDFGOPSGAURCEO-UHFFFAOYSA-N
SMILES: CCN1C(=O)C=CC1=O
Boiling Point 
  • (5)
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (8)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (4)
Physical Form 
  • (3)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (9)
  • (5)
  • (3)
  • (4)
  • (8)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)
  • (3)
  • (7)
  • (1)
  • (1)