accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 128074-72-6

IUPAC Name: 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride
Synonyms: ciprofloxacin hydrochloride|ciprofloxacin hcl|ciprofloxacin hydrochloride (anh.)|ciprofloxacin hydrochloride (anhydrous)
Molecular Weight (g/mol): 367.81
Molecular Formula: C17H19ClFN3O3
InChi Key: DIOIOSKKIYDRIQ-UHFFFAOYSA-N
SMILES: Cl.OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
Quantity 
  • (1)
  • (1)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results