accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONHN

CAS RN 128143-88-4

IUPAC Name: 6-(pyridin-2-yl)-1,4-dihydro-[2,2'-bipyridin]-4-one
Synonyms: 6-(pyridin-2-yl)-1H-[2,2'-bipyridin]-4-one
Molecular Weight (g/mol): 249.27
Molecular Formula: C15H11N3O
InChi Key: HRORSVNZQWCZTD-UHFFFAOYSA-N
SMILES: O=C1C=C(NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)