Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Bis(2-pyridyl)-4(1H)-pyridone 98.0+%, TCI America™
SDP

Catalog No. B32971G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B32971G Supplier TCI America Supplier No. B32971G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 128143-88-4
Molecular Formula C15H11N3O
Molecular Weight (g/mol) 249.273
MDL Number MFCD01321386
InChI Key HRORSVNZQWCZTD-UHFFFAOYSA-N
Synonym 2,6-Di(2-pyridyl)-4(1H)-pyridone
PubChem CID 619353
IUPAC Name 2,6-dipyridin-2-yl-1H-pyridin-4-one
SMILES C1=CC=NC(=C1)C2=CC(=O)C=C(N2)C3=CC=CC=N3

Specifications

Melting Point 178°C
Color White-Yellow
Quantity 1 g
Formula Weight 249.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(2-pyridyl)-4(1H)-pyridone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.