CAS RN 128189-46-8
IUPAC Name:
1-phenylheptadecan-1-one
Synonyms:
1-phenylheptadecan-1-one
Molecular Weight (g/mol):
330.56
Molecular Formula:
C23H38O
InChi Key:
VUXIOSYYRVXOOT-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Heptadecanophenone 98.0+%, TCI America™
CAS: 128189-46-8 Molecular Formula: C23H38O Molecular Weight (g/mol): 330.556 MDL Number: MFCD00191443 InChI Key: VUXIOSYYRVXOOT-UHFFFAOYSA-N PubChem CID: 577626 IUPAC Name: 1-phenylheptadecan-1-one SMILES: CCCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1