Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptadecanophenone 98.0+%, TCI America™
SDP

Catalog No. H078025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H078025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H078025G Supplier TCI America Supplier No. H078025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 128189-46-8
Molecular Formula C23H38O
Molecular Weight (g/mol) 330.556
MDL Number MFCD00191443
InChI Key VUXIOSYYRVXOOT-UHFFFAOYSA-N
PubChem CID 577626
IUPAC Name 1-phenylheptadecan-1-one
SMILES CCCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 57°C
Color White
Quantity 25 g
Formula Weight 330.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Heptadecanophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.