accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 128273-56-3

NH2SFFF

CAS RN 128273-56-3

IUPAC Name: 1-{4-[(trifluoromethyl)sulfanyl]phenyl}methanamine
Synonyms: 1-{4-[(trifluoromethyl)sulfanyl]phenyl}methanamine
Molecular Weight (g/mol): 207.21
Molecular Formula: C8H8F3NS
InChi Key: LACURGWEZCFLBO-UHFFFAOYSA-N
SMILES: NCC1=CC=C(SC(F)(F)F)C=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)