accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3SiH3CCH3ONN

CAS RN 128302-78-3

IUPAC Name: 2-[(tert-butyldimethylsilyl)oxy]propanedinitrile
Synonyms: 2-[(tert-butyldimethylsilyl)oxy]propanedinitrile
Molecular Weight (g/mol): 196.33
Molecular Formula: C9H16N2OSi
InChi Key: AIQDCGCDLHCZHB-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OC(C#N)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results