accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNHNOOCH3HCl

CAS RN 128322-44-1

IUPAC Name: 6-(4-methoxyphenyl)-2-methyl-3H,7H-imidazo[1,2-a]pyrazin-3-one hydrochloride
Synonyms: 6-(4-methoxyphenyl)-2-methyl-7H-imidazo[1,2-a]pyrazin-3-one hydrochloride
Molecular Weight (g/mol): 291.74
Molecular Formula: C14H14ClN3O2
InChi Key: KGMPQNFFCUDXSO-UHFFFAOYSA-N
SMILES: Cl.COC1=CC=C(C=C1)C1=CN2C(=O)C(C)=NC2=CN1
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results