Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCLA 98.0+%, TCI America™
SDP

Catalog No. A530910MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A530910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A530910MG Supplier TCI America Supplier No. A530910MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 128322-44-1
Molecular Formula C14H14ClN3O2
Molecular Weight (g/mol) 291.735
MDL Number MFCD00144728
InChI Key KGMPQNFFCUDXSO-UHFFFAOYSA-N
Synonym 2-Methyl-6-(4-methoxyphenyl)-3,7-dihydroimidazo[1,2-a]pyrazin-3-one Hydrochloride, 6-(4-Methoxyphenyl)-2-methyl-3,7-dihydroimidazo[1,2-a]pyrazin-3-one Hydrochloride
PubChem CID 16218650
IUPAC Name 6-(4-methoxyphenyl)-2-methyl-7H-imidazo[1,2-a]pyrazin-3-one;hydrochloride
SMILES CC1=NC2=CNC(=CN2C1=O)C3=CC=C(C=C3)OC.Cl

Specifications

Physical Form Powder/Crystals
Quantity 10 mg
Formula Weight 291.74
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material MCLA [Chemiluminescence Reagent]
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.