accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHO(S)SS(S)OHN(R)H3CH3COHNOO(S)NHOCH3CH3(Z)(E)

CAS RN 128517-07-7

IUPAC Name: (1S,4S,7Z,10S,16E,21R)-7-ethylidene-4,21-bis(propan-2-yl)-2-oxa-12,13-dithia-5,8,20,23-tetraazabicyclo[8.7.6]tricos-16-ene-3,6,9,19,22-pentone
Synonyms: romidepsin istodax antibiotic FR 901228 chromadax
Molecular Weight (g/mol): 540.69
Molecular Formula: C24H36N4O6S2
InChi Key: OHRURASPPZQGQM-GCCNXGTGSA-N
SMILES: C\C=C1/NC(=O)[C@H]2CSSCC\C=C\[C@H](CC(=O)N[C@H](C(C)C)C(=O)N2)OC(=O)[C@@H](NC1=O)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)